Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002092.3 | NP_002083.3 | 1, 2 | 448 | ||
GenBank mRNA | AK055499.1 | - | 2, 3 | 224 | ||
AK056891.1 | - | 2, 3 | 194 | |||
AK090755.1 | - | 2, 3 | 228 | |||
AK095799.1 | - | 2, 3 | 214 | |||
AK097055.1 | - | 2, 3 | 294 | |||
AK124124.1 | - | 2, 3 | 365 | |||
AK127161.1 | - | 2, 3 | 323 | |||
AK296287.1 | - | 2, 3 | 283 | |||
DN990192.1 | - | 1, 2 | 456 | |||
U07231.1 | - | 2, 3 | 258 |