Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014399.3 | NP_055214.1 | 6 | 1491 | ||
GenBank mRNA | AF100759.1 | - | 6 | 1412 | ||
AK026587.1 | - | 6 | 1455 | |||
AK075475.1 | - | 6 | 1436 | |||
AK093487.1 | - | 6 | 1453 | |||
AY358995.1 | - | 6 | 1422 | |||
BC033863.1 | - | 6 | 1341 | 5187772 | ||
BX647338.1 | - | 1 | 1615 | |||
CB852023.1 | - | 1 | 113 |