Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014415.3 | NP_055230.2 | 2 | 598 | ||
XM_011512689.2 | XP_011510991.1 | 2 | 306 | |||
GenBank mRNA | AK312859.1 | - | 2 | 590 | ||
BC111700.1 | - | 2 | 505 | 40038346 | ||
U69274.1 | - | 2 | 529 |