Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014415.3 | NP_055230.2 | 4 | 1201 | ||
XM_011512689.2 | XP_011510991.1 | 4 | 909 | |||
XM_011512690.2 | XP_011510992.1 | 2 | 2586 | |||
GenBank mRNA | AK312859.1 | - | 4 | 1193 | ||
BC111700.1 | - | 4 | 1108 | 40038346 | ||
U69274.1 | - | 4 | 1132 |