Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002098.5 | NP_002089.4 | 1 | 210 | ||
GenBank mRNA | BQ636025.1 | - | 1 | 187 | ||
BX537393.1 | - | 1 | 210 |