Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198182.2 | NP_937825.2 | 16 | 2428 | ||
XM_005246159.3 | XP_005246216.1 | 15 | 2314 | |||
XM_006711882.2 | XP_006711945.1 | 16 | 2464 | |||
XM_006711884.3 | XP_006711947.1 | 15 | 2343 | |||
XM_011510343.2 | XP_011508645.1 | 16 | 2435 | |||
XM_017003900.1 | XP_016859389.1 | 16 | 2537 | |||
XM_017003901.1 | XP_016859390.1 | 16 | 2501 | |||
GenBank mRNA | AF411210.1 | - | 12 | 1783 | ||
BF573452.1 | - | 204 |