Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFCP2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014553.2 | NP_055368.1 | 12 | 1204 | ||
XM_017003902.1 | XP_016859391.1 | 12 | 1212 | |||
XM_017003903.1 | XP_016859392.1 | 7 | 511 | |||
GenBank mRNA | AF198488.1 | - | 12 | 1201 | ||
AL137740.1 | - | 12 | 1203 | |||
BC064698.1 | - | 12 | 1204 | 30347863 |