Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201457.1 | NP_001188386.1 | 14 | 1844 | ||
NM_031272.4 | NP_112562.3 | 14 | 1826 | |||
NM_198393.3 | NP_938207.2 | 14 | 1826 | |||
XM_011525029.2 | XP_011523331.1 | 14 | 1805 | |||
XM_017024860.1 | XP_016880349.1 | 12 | 1476 | |||
XM_017024861.1 | XP_016880350.1 | 13 | 1651 | |||
GenBank mRNA | AF285601.1 | - | 3 | 290 | ||
AK128217.1 | - | 10 | 1261 | |||
AK308299.1 | - | 14 | 1786 | |||
AL834143.1 | - | 14 | 1825 | |||
BC040526.1 | - | 14 | 1812 | 4831308 |