Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161587.1 | NP_001155059.1 | 2 | 515 | ||
NM_002103.4 | NP_002094.2 | 2 | 515 | |||
GenBank mRNA | AK223579.1 | - | 2 | 471 | ||
AK291672.1 | - | 2 | 471 | |||
AK300369.1 | - | 2 | 303 | |||
BC002617.2 | - | 2 | 454 | 3143019 | ||
BC003182.1 | - | 2 | 472 | 2958297 | ||
BC007688.2 | - | 2 | 441 | 3953692 | ||
J04501.1 | - | 2 | 434 | |||
U32573.1 | - | 2 | 416 |