Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC40A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014585.5 | NP_055400.1 | 8 | 2850 | ||
XM_005246505.1 | XP_005246562.1 | 9 | 2985 | |||
XM_017003938.1 | XP_016859427.1 | 9 | 3099 | |||
GenBank mRNA | AF215636.1 | - | 8 | 2804 | ||
AK002038.1 | - | 3 | 1783 | |||
AK223236.1 | - | 8 | 2825 | |||
AL136944.1 | - | 8 | 2813 | |||
BC037733.1 | - | 8 | 2929 | 4823308 | ||
BX537503.1 | - | 1 | 2680 |