Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHCR24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014762.3 | NP_055577.1 | 9 | 2009 | ||
GenBank mRNA | AB073391.1 | - | 10 | 1884 | ||
AF261758.1 | - | 9 | 1979 | |||
AK304302.1 | - | 6 | 1297 | |||
AK304421.1 | - | 5 | 1136 | |||
AK316427.1 | - | 9 | 1710 | |||
BC004375.1 | - | 9 | 1915 | 3638639 | ||
BC011669.2 | - | 9 | 1942 | 4130396 | ||
D13643.3 | - | 9 | 1992 |