Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SV2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167580.1 | NP_001161052.1 | 9 | 1379 | ||
NM_001323031.1 | NP_001309960.1 | 10 | 1912 | |||
NM_001323032.1 | NP_001309961.1 | 10 | 2221 | |||
NM_001323033.1 | NP_001309962.1 | 8 | 1888 | |||
NM_001323034.1 | NP_001309963.1 | 10 | 2221 | |||
NM_001323036.1 | NP_001309965.1 | 9 | 1198 | |||
NM_001323037.1 | NP_001309966.1 | 11 | 2128 | |||
NM_001323038.1 | NP_001309967.1 | 10 | 2040 | |||
NM_001323039.1 | NP_001309968.1 | 10 | 2313 | |||
NM_001323040.1 | NP_001309969.1 | 10 | 1559 | |||
NM_014848.5 | NP_055663.1 | 11 | 2309 | |||
XM_005254998.3 | XP_005255055.1 | 10 | 1974 | |||
XM_017022761.1 | XP_016878250.1 | 10 | 1925 | |||
XM_017022762.1 | XP_016878251.1 | 10 | 2060 | |||
GenBank mRNA | AB018278.1 | - | 9 | 1582 | ||
AK294902.1 | - | 9 | 1381 | |||
BC030011.2 | - | 11 | 1946 | 4826206 | ||
CR457086.1 | - | 9 | 1418 |