Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258040.2 | NP_001244969.1 | 2 | 533 | ||
NM_001258041.2 | NP_001244970.1 | 2 | 533 | |||
NM_001258042.2 | NP_001244971.1 | 2 | 533 | |||
NM_001289092.1 | NP_001276021.1 | 2 | 533 | |||
NM_001289093.1 | NP_001276022.1 | 2 | 533 | |||
NM_001289094.1 | NP_001276023.1 | 2 | 446 | |||
NM_002109.5 | NP_002100.2 | 2 | 533 | |||
GenBank mRNA | AK000498.1 | - | 2 | 213 | ||
AK055917.1 | - | 2 | 147 | |||
AK225776.1 | - | 2 | 183 | |||
AK293154.1 | - | 2 | 147 | |||
AK293531.1 | - | 195 | ||||
AK295219.1 | - | 2 | 189 | |||
AK302295.1 | - | 2 | 147 | |||
AK303778.1 | - | 2 | 343 | |||
BC011807.2 | - | 2 | 140 | 4331277 | ||
BC080514.1 | - | 2 | 138 | 2821065 | ||
DC394573.1 | - | 2 | 533 | |||
X05345.1 | - | 2 | 187 | |||
Z11518.1 | - | 2 | 668 |