Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282550.1 | NP_001269479.1 | 5 | 3615 | ||
NM_001282551.1 | NP_001269480.1 | 5 | 3711 | |||
NM_001282552.2 | NP_001269481.1 | 4 | 3705 | |||
NM_001282554.1 | NP_001269483.1 | 4 | 3620 | |||
NM_014962.3 | NP_055777.1 | 4 | 4033 | |||
NM_181443.3 | NP_852108.2 | 5 | 3796 | |||
XM_017027728.1 | XP_016883217.1 | 6 | 3944 | |||
XM_017027729.1 | XP_016883218.1 | 6 | 3876 | |||
XM_017027730.1 | XP_016883219.1 | 3 | 3964 | |||
GenBank mRNA | AB023169.1 | - | 4 | 3981 | ||
AK024349.1 | - | 1 | 1678 | |||
AL137756.1 | - | 1 | 4785 |