Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KDM4C SIRNA Assays ›
Gene Symbol:
KDM4C
Gene Name:
lysine demethylase 4C
Gene Aliases:
GASC1, JHDM3C, JMJD2C, TDRD14C
Chromosome Location:
Chr.9: 6720863 - 7175648 on Build GRCh38
UniGene ID:
Hs.709425
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108665
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KDM4C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146695.1
NP_001140167.1
10, 11
1942
NM_001146696.1
NP_001140168.1
10, 11
1514
NM_001304339.1
NP_001291268.1
10, 11
1942
NM_001304340.1
NP_001291269.1
9, 10
1766
NM_015061.3
NP_055876.2
10, 11
1942
NR_130707.1
12
2885
XM_006716741.2
XP_006716804.1
10, 11
1611
XM_011517811.2
XP_011516113.1
11, 12
1566
XM_011517812.2
XP_011516114.2
6, 7
756
XM_011517816.2
XP_011516118.1
3, 4
567
XM_017014498.1
XP_016869987.1
10, 11
1611
XM_017014499.1
XP_016869988.1
12, 13
1653
XM_017014500.1
XP_016869989.1
12, 13
1680
XM_017014501.1
XP_016869990.1
9, 10
1295
XM_017014502.1
XP_016869991.1
10, 11
1611
XM_017014503.1
XP_016869992.1
10, 11
1611
XM_017014504.1
XP_016869993.1
10, 11
1611
XM_017014505.1
XP_016869994.1
2, 3
297
XM_017014506.1
XP_016869995.1
11, 12
1626
GenBank mRNA
AB018323.1
-
10, 11
1522
AB037901.1
-
10, 11
1512
AK296401.1
-
3, 4
612
AK303651.1
-
9, 10
1352
AK304032.1
-
10, 11
1440
AK304095.1
-
9, 10
1339
AK310439.1
-
10, 11
1525
AK314355.1
-
10, 11
1507
BC104859.1
-
10, 11
1422
8143862
BC104861.1
-
10, 11
1422
8143864
BC143571.1
-
10, 11
1422
9052082
Back To Top
More Information
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
positive regulation of cell proliferation
histone H3-K9 demethylation
oxidation-reduction process
zinc ion binding
enzyme binding
histone demethylase activity
histone demethylase activity (H3-K9 specific)
androgen receptor binding
dioxygenase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline