Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271620.2 | NP_001258549.1 | 8 | 4083 | ||
NM_015069.4 | NP_055884.2 | 8 | 3973 | |||
XM_005255856.4 | XP_005255913.1 | 8 | 4686 | |||
XM_005255857.4 | XP_005255914.1 | 6 | 4176 | |||
XM_006721171.3 | XP_006721234.1 | 11 | 4415 | |||
XM_017023076.1 | XP_016878565.1 | 8 | 4700 | |||
XM_017023077.1 | XP_016878566.1 | 8 | 4049 | |||
XM_017023078.1 | XP_016878567.1 | 7 | 4022 | |||
GenBank mRNA | AB018303.2 | - | 6 | 3740 | ||
AK027479.1 | - | 5 | 2139 | |||
AK304829.1 | - | 6 | 3777 | |||
AK309835.1 | - | 6 | 3758 |