Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SATB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172509.1 | NP_001165980.1 | 7 | 1748 | ||
NM_001172517.1 | NP_001165988.1 | 8 | 1344 | |||
NM_015265.3 | NP_056080.1 | 8 | 1324 | |||
XM_005246396.2 | XP_005246453.1 | 6 | 979 | |||
XM_006712372.2 | XP_006712435.1 | 7 | 1094 | |||
XM_011510840.2 | XP_011509142.1 | 8 | 1318 | |||
XM_017003656.1 | XP_016859145.1 | 7 | 979 | |||
GenBank mRNA | AB028957.1 | - | 7 | 1017 | ||
AB209376.1 | - | 8 | 1344 | |||
AK025127.1 | - | 1 | 234 | |||
AK056638.1 | - | 7 | 1749 | |||
AK291463.1 | - | 8 | 1330 | |||
BC098136.1 | - | 6 | 982 | 40007830 | ||
BC099723.1 | - | 6 | 982 | 40007832 | ||
BC103492.1 | - | 3 | 628 | 40007829 | ||
BC103500.1 | - | 6 | 982 | 40007831 |