Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278926.1 | NP_001265855.1 | 6 | 957 | ||
NM_001278927.1 | NP_001265856.1 | 5 | 747 | |||
NM_001278929.1 | NP_001265858.1 | 4 | 817 | |||
NM_015342.3 | NP_056157.1 | 5 | 923 | |||
XM_006714583.3 | XP_006714646.1 | 6 | 893 | |||
XM_011543295.1 | XP_011541597.1 | 4 | 1205 | |||
GenBank mRNA | AK025679.1 | - | 5 | 870 | ||
AK298175.1 | - | 5 | 687 | |||
AK300022.1 | - | 6 | 900 | |||
AK301654.1 | - | 4 | 757 | |||
BC015385.1 | - | 5 | 851 | 4390706 | ||
D38552.1 | - | 5 | 846 |