Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244883.1 | NP_001231812.1 | 4, 5 | 272 | ||
NM_001933.4 | NP_001924.2 | 4, 5 | 272 | |||
NR_045209.1 | 3, 4 | 223 | ||||
GenBank mRNA | AK289414.1 | - | 4, 5 | 270 | ||
AK300396.1 | - | 4, 5 | 241 | |||
AU133381.1 | - | 4, 5 | 247 | |||
BC000302.2 | - | 4, 5 | 234 | 2820348 | ||
BC001922.1 | - | 4, 5 | 234 | 2820348 | ||
BX248774.1 | - | 3, 4 | 158 | |||
CR456727.1 | - | 4, 5 | 209 | |||
D16373.1 | - | 4, 5 | 221 | |||
DC338747.1 | - | 4, 5 | 273 | |||
L37418.1 | - | 4, 5 | 209 |