Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001935.3 | NP_001926.2 | 6 | 943 | ||
XM_005246371.2 | XP_005246428.1 | 6 | 520 | |||
GenBank mRNA | AB451339.1 | - | 5 | 382 | ||
AB451488.1 | - | 5 | 382 | |||
AK314798.1 | - | 6 | 520 | |||
AY429532.1 | - | 6 | 477 | |||
BC013329.2 | - | 5 | 377 | |||
BC065265.1 | - | 6 | 943 | 6156880 | ||
M74777.1 | - | 6 | 392 | |||
M80536.1 | - | 5 | 382 | |||
X60708.1 | - | 6 | 457 |