Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130110.1 | NP_001123582.1 | 2 | 823 | ||
NM_015559.2 | NP_056374.2 | 2 | 724 | |||
GenBank mRNA | AB007897.2 | - | 2 | 724 | ||
AB022660.1 | - | 1 | 271 | |||
BC062338.1 | - | 2 | 823 | |||
BC146776.1 | - | 2 | 724 | 9007225 |