Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199917.1 | NP_001186846.1 | 4 | 525 | ||
NM_001199918.1 | NP_001186847.1 | 2 | 183 | |||
NM_001199919.1 | NP_001186848.1 | 3 | 389 | |||
NM_015599.2 | NP_056414.1 | 3 | 389 | |||
XM_017010935.1 | XP_016866424.1 | 2 | 156 | |||
XM_017010936.1 | XP_016866425.1 | 3 | 388 | |||
XM_017010937.1 | XP_016866426.1 | 2 | 156 | |||
XM_017010938.1 | XP_016866427.1 | 3 | 377 | |||
GenBank mRNA | AB032081.1 | - | 3 | 301 | ||
AF102265.1 | - | 3 | 305 | |||
AF180371.1 | - | 3 | 312 | |||
AK001365.1 | - | 2 | 124 | |||
AK023432.1 | - | 3 | 334 | |||
AK225363.1 | - | 3 | 327 | |||
AK301867.1 | - | 4 | 503 | |||
AK314512.1 | - | 3 | 322 | |||
BC001258.1 | - | 3 | 348 | 3451677 | ||
DB004046.1 | - | 3 | 390 | |||
DB033430.1 | - | 3 | 388 |