Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164342.2 | NP_001157814.1 | 5 | 2083 | ||
NM_001164343.2 | NP_001157815.1 | 12 | 2545 | |||
NM_001164344.2 | NP_001157816.1 | 8 | 2509 | |||
NM_001164345.2 | NP_001157817.1 | 7 | 2463 | |||
NM_001164346.2 | NP_001157818.1 | 6 | 2102 | |||
NM_001164347.2 | NP_001157819.1 | 6 | 2124 | |||
NM_015642.5 | NP_056457.3 | 10 | 2307 | |||
NR_121662.1 | 7 | 565 | ||||
GenBank mRNA | AF139460.2 | - | 5 | 2089 | ||
AK129968.1 | - | 1 | 626 | |||
AK225788.1 | - | 7 | 2459 | |||
AK290116.1 | - | 3 | 1845 | |||
AK315307.1 | - | 6 | 2102 | |||
AL050276.1 | - | 8 | 2172 | |||
BC029041.1 | - | 6 | 2124 | 5184140 | ||
BC090936.1 | - | 12 | 2506 | |||
BX647778.1 | - | 10 | 2308 | |||
CB164748.1 | - | 6 | 550 |