Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290261.1 | NP_001277190.1 | 4 | 2352 | ||
NM_015655.3 | NP_056470.1 | 5 | 2025 | |||
XM_005260702.3 | XP_005260759.1 | 4 | 1841 | |||
XM_006723558.3 | XP_006723621.1 | 5 | 3028 | |||
XM_011529219.2 | XP_011527521.1 | 5 | 2148 | |||
XM_017027802.1 | XP_016883291.1 | 3 | 3923 | |||
XM_017027803.1 | XP_016883292.1 | 4 | 2932 | |||
XM_017027804.1 | XP_016883293.1 | 2 | 1701 | |||
GenBank mRNA | AK056184.1 | - | 5 | 1936 | ||
AK056466.1 | - | 1 | 1548 | |||
AK291235.1 | - | 5 | 1962 | |||
AK299811.1 | - | 3 | 2027 | |||
AL049942.1 | - | 1 | 1066 | |||
BC021298.2 | - | 5 | 1954 | 4551314 |