Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015906.3 | NP_056990.3 | 12, 13 | 2286 | ||
NM_033020.2 | NP_148980.2 | 12, 13 | 2286 | |||
XM_005270936.3 | XP_005270993.1 | 13, 14 | 2279 | |||
XM_005270937.3 | XP_005270994.1 | 13, 14 | 2279 | |||
XM_011541568.2 | XP_011539870.1 | 13, 14 | 2276 | |||
XM_017001452.1 | XP_016856941.1 | 10, 11 | 1561 | |||
XM_017001453.1 | XP_016856942.1 | 10, 11 | 1561 | |||
GenBank mRNA | AB029036.1 | - | 12, 13 | 2215 | ||
AF119043.1 | - | 12, 13 | 2286 | |||
AF220136.1 | - | 12, 13 | 2286 | |||
AF220137.1 | - | 12, 13 | 2286 | |||
AJ132948.1 | - | 12, 13 | 1977 | |||
AK023434.1 | - | 10, 11 | 1499 | |||
AK057487.1 | - | 7, 8 | 873 | |||
AK301997.1 | - | 7, 8 | 964 |