Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015906.3 | NP_056990.3 | 20 | 3380 | ||
NM_033020.2 | NP_148980.2 | 19 | 3329 | |||
XM_005270936.3 | XP_005270993.1 | 21 | 3373 | |||
XM_005270937.3 | XP_005270994.1 | 20 | 3322 | |||
XM_011541568.2 | XP_011539870.1 | 21 | 3370 | |||
XM_017001452.1 | XP_016856941.1 | 18 | 2655 | |||
XM_017001453.1 | XP_016856942.1 | 17 | 2604 | |||
GenBank mRNA | AB029036.1 | - | 20 | 3309 | ||
AF119043.1 | - | 20 | 3380 | |||
AF220136.1 | - | 20 | 3380 | |||
AF220137.1 | - | 19 | 3329 | |||
AJ132948.1 | - | 20 | 3071 | |||
AK023434.1 | - | 18 | 2593 | |||
AK057487.1 | - | 15 | 1597 | |||
AK301997.1 | - | 15 | 2058 |