Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSDHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129765.1 | NP_001123237.1 | 9 | 1547 | ||
NM_015922.2 | NP_057006.1 | 8 | 1480 | |||
XM_011531178.2 | XP_011529480.1 | 10 | 1607 | |||
XM_017029564.1 | XP_016885053.1 | 8 | 1412 | |||
GenBank mRNA | AK026549.1 | - | 8 | 1370 | ||
BC007816.2 | - | 8 | 1451 | 4299819 | ||
U47105.2 | - | 8 | 1480 |