Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142541.2 | NP_001136013.1 | 11 | 1625 | ||
NM_015948.4 | NP_057032.2 | 11 | 1987 | |||
NR_109913.1 | 12 | 1978 | ||||
NR_109914.1 | 9 | 1921 | ||||
XM_005249156.3 | XP_005249213.1 | 10 | 1767 | |||
XM_005249157.2 | XP_005249214.1 | 11 | 1760 | |||
XM_005249158.1 | XP_005249215.1 | 10 | 1655 | |||
XM_006715102.3 | XP_006715165.1 | 11 | 1797 | |||
XM_006715103.3 | XP_006715166.1 | 9 | 1674 | |||
XM_011514648.1 | XP_011512950.1 | 12 | 1767 | |||
XM_017010910.1 | XP_016866399.1 | 11 | 2083 | |||
XM_017010911.1 | XP_016866400.1 | 10 | 2001 | |||
XM_017010912.1 | XP_016866401.1 | 10 | 1622 | |||
XM_017010913.1 | XP_016866402.1 | 10 | 1990 | |||
XM_017010914.1 | XP_016866403.1 | 10 | 1851 | |||
XM_017010915.1 | XP_016866404.1 | 9 | 1562 | |||
GenBank mRNA | BC006973.1 | - | 11 | 1813 | 3827889 | |
BX538271.1 | - | 12 | 1974 | |||
BX641086.1 | - | 10 | 7126 | |||
BX647208.1 | - | 9 | 1855 | |||
BX648022.1 | - | 10 | 5924 | |||
DB576665.1 | - | 1 | 243 |