Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF589 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016089.2 | NP_057173.2 | 4 | 2438 | ||
XM_017006575.1 | XP_016862064.1 | 4 | 2519 | |||
XM_017006576.1 | XP_016862065.1 | 6 | 2668 | |||
XM_017006577.1 | XP_016862066.1 | 4 | 3724 | |||
XM_017006578.1 | XP_016862067.1 | 4 | 2499 | |||
XM_017006579.1 | XP_016862068.1 | 6 | 2624 | |||
GenBank mRNA | AB209387.1 | - | 4 | 2419 | ||
AF114816.1 | - | 7 | 1457 | |||
AF114817.1 | - | 6 | 2741 | |||
AL832831.1 | - | 4 | 2425 | |||
AY258146.1 | - | 4 | 2438 | |||
BC005859.2 | - | 4 | 2397 | |||
BC006247.2 | - | 4 | 2418 | |||
BC008213.1 | - | 1 | 1006 | |||
BC048798.1 | - | 4 | 2415 |