Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016120.3 | NP_057204.2 | 4 | 1320 | ||
NM_183353.2 | NP_899196.1 | 5 | 1382 | |||
GenBank mRNA | AF155109.1 | - | 4 | 1272 | ||
AK001334.1 | - | 4 | 1302 | |||
AK314760.1 | - | 1 | 876 | |||
BC013357.2 | - | 4 | 1284 | 3834870 |