Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016205.2 | NP_057289.1 | 3 | 962 | ||
NR_036641.1 | 4 | 1023 | ||||
XM_011532124.2 | XP_011530426.1 | 5 | 642 | |||
XM_017008455.1 | XP_016863944.1 | 3 | 591 | |||
XM_017008456.1 | XP_016863945.1 | 3 | 924 | |||
GenBank mRNA | AB033831.1 | - | 3 | 797 | ||
AF091434.1 | - | 3 | 962 | |||
AF244813.1 | - | 3 | 659 | |||
AF260738.1 | - | 3 | 633 | |||
AK300480.1 | - | 2 | 448 | |||
AK313817.1 | - | 3 | 692 | |||
AM922296.1 | - | 4 | 843 | |||
AY358493.1 | - | 3 | 755 | |||
BC094746.1 | - | 3 | 563 | |||
BC136662.1 | - | 3 | 883 | 9020652 |