Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016205.2 | NP_057289.1 | 5 | 1316 | ||
NR_036641.1 | 6 | 1377 | ||||
XM_011532124.2 | XP_011530426.1 | 7 | 996 | |||
XM_017008455.1 | XP_016863944.1 | 5 | 945 | |||
GenBank mRNA | AB033831.1 | - | 5 | 1151 | ||
AF091434.1 | - | 5 | 1316 | |||
AF244813.1 | - | 5 | 1013 | |||
AF260738.1 | - | 5 | 987 | |||
AK300480.1 | - | 4 | 802 | |||
AK304638.1 | - | 7 | 659 | |||
AK313817.1 | - | 5 | 1046 | |||
AM922296.1 | - | 6 | 1197 | |||
AY358493.1 | - | 5 | 1109 | |||
BC094746.1 | - | 5 | 917 | |||
BC136662.1 | - | 5 | 1237 | 9020652 |