Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016322.3 | NP_057406.2 | 8 | 1291 | ||
GenBank mRNA | AK023524.1 | - | 8 | 1073 | ||
AK090889.1 | - | 8 | 1090 | |||
AL162081.1 | - | 8 | 1018 |