Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2IRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199207.1 | NP_001186136.1 | 10, 11 | 1797 | ||
NM_005685.3 | NP_005676.3 | 10, 11 | 1701 | |||
NM_016328.2 | NP_057412.1 | 10, 11 | 1701 | |||
XM_006716182.3 | XP_006716245.1 | 10, 11 | 1497 | |||
XM_006716183.3 | XP_006716246.1 | 10, 11 | 1342 | |||
XM_011516713.1 | XP_011515015.1 | 10, 11 | 1497 | |||
XM_017012802.1 | XP_016868291.1 | 10, 11 | 1497 | |||
XM_017012803.1 | XP_016868292.1 | 10, 11 | 1497 | |||
XM_017012804.1 | XP_016868293.1 | 10, 11 | 1426 | |||
XM_017012805.1 | XP_016868294.1 | 10, 11 | 1497 | |||
GenBank mRNA | AB209389.1 | - | 10, 11 | 2999 | ||
AF089107.1 | - | 9, 10 | 1315 | |||
AF104923.1 | - | 10, 11 | 1347 | |||
AF118270.2 | - | 10, 11 | 1356 | |||
AF151354.1 | - | 10, 11 | 1701 | |||
AF156489.1 | - | 10, 11 | 1404 | |||
AK308242.1 | - | 4, 5 | 369 | |||
AY648295.1 | - | 10, 11 | 1617 | |||
BC018136.1 | - | 10, 11 | 1466 | 3913745 |