Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001962.2 | NP_001953.1 | 2 | 682 | ||
XM_006714565.2 | XP_006714628.1 | 2 | 2151 | |||
XM_011543250.2 | XP_011541552.1 | 2 | 495 | |||
XM_011543251.2 | XP_011541553.1 | 2 | 2008 | |||
XM_017009205.1 | XP_016864694.1 | 2 | 1771 | |||
GenBank mRNA | AK308672.1 | - | 2 | 672 | ||
BC075054.2 | - | 2 | 449 | 30915365 | ||
BC075055.2 | - | 2 | 449 | 30915581 | ||
U26403.1 | - | 2 | 682 |