Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130678.1 | NP_001124150.1 | 4, 5 | 473 | ||
NM_001130679.1 | NP_001124151.1 | 4, 5 | 1816 | |||
NM_001968.3 | NP_001959.1 | 4, 5 | 1816 | |||
XM_006714126.3 | XP_006714189.1 | 5, 6 | 1519 | |||
GenBank mRNA | AK295083.1 | - | 4, 5 | 309 | ||
AK300982.1 | - | 4, 5 | 473 | |||
AV708571.1 | - | 4, 5 | 389 | |||
BC012611.1 | - | 4, 5 | 309 | 4282826 | ||
BC035166.1 | - | 4, 5 | 1824 | 5265600 | ||
BC043226.1 | - | 4, 5 | 1155 | 5295521 | ||
BC107715.1 | - | 4, 5 | 310 | 6716042 | ||
BM849222.1 | - | 4, 5 | 307 | |||
DC388666.1 | - | 4, 5 | 473 | |||
KJ534830.1 | - | 4, 5 | 353 | |||
M15353.1 | - | 4, 5 | 311 |