Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016614.2 | NP_057698.2 | 7 | 1317 | ||
GenBank mRNA | AF223469.1 | - | 7 | 1308 | ||
AJ269473.1 | - | 7 | 1313 | |||
AJ420495.1 | - | 6 | 1518 | |||
AK002168.1 | - | 7 | 1292 | |||
AK296623.1 | - | 6 | 993 | |||
AK298699.1 | - | 7 | 1522 | |||
BC017553.2 | - | 7 | 1329 | 3920790 | ||
BC110375.1 | - | 7 | 1298 | 5737150 |