Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016614.2 | NP_057698.2 | 7 | 1445 | ||
GenBank mRNA | AF223469.1 | - | 7 | 1436 | ||
AJ269473.1 | - | 7 | 1441 | |||
AJ420495.1 | - | 6 | 1646 | |||
AK002168.1 | - | 7 | 1420 | |||
AK298699.1 | - | 7 | 1650 | |||
BC017553.2 | - | 7 | 1457 | 3920790 | ||
BC110375.1 | - | 7 | 1426 | 5737150 |