Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAXBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013329.3 | NP_037461.2 | 7 | 1472 | ||
NM_016631.3 | NP_057715.2 | 7 | 1472 | |||
NR_027873.1 | 7 | 1472 | ||||
XM_006724066.1 | XP_006724129.1 | 7 | 1401 | |||
XM_011529804.2 | XP_011528106.1 | 7 | 1401 | |||
GenBank mRNA | AF231920.1 | - | 6 | 889 | ||
AJ279080.1 | - | 7 | 1298 | |||
AJ279081.1 | - | 7 | 1298 | |||
AY033903.1 | - | 7 | 1282 | |||
AY033904.1 | - | 7 | 1282 | |||
AY033905.1 | - | 7 | 1282 | |||
AY033906.1 | - | 7 | 1282 |