Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017429.2 | NP_059125.2 | 11 | 2334 | ||
XM_011523109.1 | XP_011521411.1 | 9 | 2182 | |||
XM_017023286.1 | XP_016878775.1 | 9 | 2176 | |||
XM_017023287.1 | XP_016878776.1 | 10 | 2271 | |||
XM_017023289.1 | XP_016878778.1 | 7 | 1448 | |||
GenBank mRNA | AF294900.1 | - | 11 | 2334 | ||
AK001592.1 | - | 11 | 2334 |