Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017514.4 | NP_059984.3 | 17 | 3132 | ||
XM_005274706.4 | XP_005274763.2 | 17 | 2804 | |||
XM_006724829.3 | XP_006724892.1 | 17 | 3036 | |||
XM_011531183.2 | XP_011529485.1 | 16 | 2496 | |||
GenBank mRNA | BQ718472.1 | - | 4 | 327 | ||
CD628272.1 | - | 4 | 369 | |||
X87852.1 | - | 17 | 3141 |