Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017564.9 | NP_060034.9 | 69 | 8195 | ||
XM_011538539.2 | XP_011536841.1 | 55 | 6439 | |||
XM_011538541.2 | XP_011536843.1 | 47 | 5546 | |||
XM_011538542.2 | XP_011536844.1 | 36 | 4355 | |||
GenBank mRNA | AB052958.1 | - | 69 | 8191 | ||
AF160476.1 | - | 24 | 3217 | |||
AJ295695.1 | - | 69 | 8195 | |||
AK024503.1 | - | 37 | 4537 | |||
AK074051.1 | - | 47 | 5566 | |||
AK160380.1 | - | 68 | 7913 | |||
AL133021.1 | - | 28 | 3570 | |||
AY311388.1 | - | 69 | 8213 |