Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320747.1 | NP_001307676.1 | 3, 4 | 601 | ||
NM_001988.3 | NP_001979.2 | 3, 4 | 601 | |||
GenBank mRNA | AK290333.1 | - | 3, 4 | 601 | ||
BC126103.1 | - | 3, 4 | 380 | 8991819 | ||
BC126105.1 | - | 3, 4 | 380 | 8991821 | ||
BC143814.1 | - | 3, 4 | 380 | 9052329 | ||
U53786.1 | - | 3, 4 | 445 |