Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193513.1 | NP_001180442.1 | 13, 14 | 1004 | ||
NM_001193515.1 | NP_001180444.1 | 11, 12 | 797 | |||
NM_017964.3 | NP_060434.2 | 12, 13 | 884 | |||
XM_006712048.3 | XP_006712111.1 | 9, 10 | 666 | |||
XM_011532960.2 | XP_011531262.1 | 12, 13 | 1140 | |||
XM_011532961.2 | XP_011531263.1 | 11, 12 | 818 | |||
XM_011532962.2 | XP_011531264.1 | 10, 11 | 969 | |||
XM_011532963.2 | XP_011531265.1 | 13, 14 | 981 | |||
XM_017004462.1 | XP_016859951.1 | 10, 11 | 731 | |||
XM_017004463.1 | XP_016859952.1 | 13, 14 | 981 | |||
GenBank mRNA | AK055663.1 | - | 12, 13 | 861 | ||
AK096672.1 | - | 11, 12 | 844 | |||
AK127716.1 | - | 8, 9 | 618 | |||
AK304225.1 | - | 11, 12 | 774 | |||
AL832755.1 | - | 9, 10 | 641 | |||
BC032525.1 | - | 3, 4 | 235 | |||
BC066903.1 | - | 13, 14 | 981 | 4801303 | ||
EF560726.1 | - | 12, 13 | 862 |