Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193513.1 | NP_001180442.1 | 15 | 1187 | ||
NM_001193514.1 | NP_001180443.1 | 13 | 998 | |||
NM_001193515.1 | NP_001180444.1 | 13 | 980 | |||
NM_017964.3 | NP_060434.2 | 14 | 1067 | |||
XM_006712048.3 | XP_006712111.1 | 11 | 849 | |||
XM_011532959.2 | XP_011531261.1 | 14 | 1095 | |||
XM_011532960.2 | XP_011531262.1 | 14 | 1323 | |||
XM_011532961.2 | XP_011531263.1 | 13 | 1001 | |||
XM_011532962.2 | XP_011531264.1 | 12 | 1152 | |||
XM_017004462.1 | XP_016859951.1 | 12 | 914 | |||
GenBank mRNA | AK055663.1 | - | 14 | 1044 | ||
AK096672.1 | - | 13 | 1027 | |||
AK127716.1 | - | 10 | 801 | |||
AK304225.1 | - | 13 | 957 | |||
AL832755.1 | - | 11 | 824 | |||
BC032525.1 | - | 5 | 418 | |||
BC066903.1 | - | 15 | 1164 | 4801303 | ||
EF560719.1 | - | 13 | 976 | |||
EF560726.1 | - | 14 | 1045 |