Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018010.3 | NP_060480.1 | 5 | 892 | ||
GenBank mRNA | AB014762.1 | - | 5 | 644 | ||
AF139576.1 | - | 5 | 722 | |||
AF245220.1 | - | 5 | 786 | |||
AK001009.1 | - | 5 | 731 | |||
AK298854.1 | - | 5 | 790 | |||
BC009690.2 | - | 1 | 59 | |||
BC011899.2 | - | 5 | 741 | 4053749 | ||
CR457251.1 | - | 5 | 644 |