Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018039.2 | NP_060509.2 | 3 | 2656 | ||
GenBank mRNA | AK001113.1 | - | 1 | 1427 | ||
AK056162.1 | - | 1 | 2068 | |||
AL137545.1 | - | 1 | 644 | |||
BC119010.1 | - | 1 | 2025 | 40085404 | ||
BC122858.1 | - | 1 | 2025 | 40085403 |