Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMG20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304504.1 | NP_001291433.1 | 3 | 538 | ||
NM_001304505.1 | NP_001291434.1 | 3 | 538 | |||
NM_018200.3 | NP_060670.1 | 4 | 631 | |||
XM_011521158.2 | XP_011519460.1 | 3 | 494 | |||
GenBank mRNA | AF146222.1 | - | 3 | 271 | ||
AK001601.1 | - | 3 | 301 | |||
AK223100.1 | - | 3 | 297 | |||
AK297449.1 | - | 2 | 231 | |||
AL355737.1 | - | 2 | 193 | |||
BC021959.1 | - | 4 | 381 | 3908842 | ||
BT006716.1 | - | 2 | 180 | |||
DA333471.1 | - | 3 | 538 | |||
DB068283.1 | - | 3 | 285 |