Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMG20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304504.1 | NP_001291433.1 | 4 | 691 | ||
NM_018200.3 | NP_060670.1 | 5 | 784 | |||
XM_011521158.2 | XP_011519460.1 | 4 | 647 | |||
GenBank mRNA | AF146222.1 | - | 4 | 424 | ||
AK001601.1 | - | 4 | 454 | |||
AK096546.1 | - | 4 | 385 | |||
AK223100.1 | - | 4 | 450 | |||
AK297449.1 | - | 3 | 384 | |||
AK316281.1 | - | 1 | 59 | |||
AL355737.1 | - | 3 | 346 | |||
BC021959.1 | - | 5 | 534 | 3908842 | ||
BT006716.1 | - | 3 | 333 |