Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGDHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143996.1 | NP_001137468.1 | 13 | 1821 | ||
NM_001143997.1 | NP_001137469.1 | 12 | 1616 | |||
NM_018245.2 | NP_060715.2 | 14 | 1992 | |||
XM_011539946.1 | XP_011538248.1 | 14 | 1877 | |||
XM_011539947.1 | XP_011538249.1 | 14 | 2083 | |||
XM_017016401.1 | XP_016871890.1 | 14 | 2267 | |||
XM_017016402.1 | XP_016871891.1 | 14 | 1894 | |||
XM_017016403.1 | XP_016871892.1 | 12 | 1640 | |||
GenBank mRNA | AB033116.3 | - | 13 | 1855 | ||
AK001713.1 | - | 14 | 1936 | |||
AK290226.1 | - | 13 | 1965 | |||
AK296551.1 | - | 13 | 1730 | |||
AK303729.1 | - | 12 | 1563 | |||
AK307759.1 | - | 13 | 1750 | |||
BC026320.1 | - | 14 | 1911 | 4797814 | ||
BP209348.1 | - | 2 | 218 |